missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Polypropylene Carboy with Silicone-Lined Cap and No Spigot
Description
- Durable polypropylene construction: Built from virgin-grade polypropylene with silicone-lined cap for long-lasting use in liquid storage and transportation.
- Autoclavable and temperature resistant: Withstands steam sterilization and temperatures from 0 to 135°C, ideal for scientific and lab applications.
- FDA and USP compliant: FDA 21 CFR 177.1520 and USP 88 Class VI certified for safe use in food, drug, and medical settings.
- Leakproof and shatter resistant: Tested for crack resistance, handle strength, and leak prevention for worry-free handling of liquids.
- Versatile use cases: Perfect for reagent storage, mixing solutions, and transporting liquids in labs, research facilities, and industrial environments.
- Non-sterile.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 25 L |
| Color | Translucent |
| Material | Polypropylene |
| Mouth Style | Narrow |
| Product Type | Carboy |
| Sterility | Non-sterile |