missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Polypropylene, Carboy with Sanitary Neck
For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Color: White (Translucent)
Supplier: Justrite™ 12936
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
| ±5% | |
| 5.3 gal. | |
| White (Translucent) | |
| For storing and dispensing solvents, buffers, reagents and powders. | |
| 13.6 in. | |
| Threadless | |
| Rectangular |
| Autoclavable | |
| 20 L | |
| 345 x 206 x 605 mm | |
| 23.8 in. | |
| Polypropylene | |
| Carboy with Sanitary Neck | |
| 8.1 in. |