missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Polypropylene, Carboy
For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White
Supplier: Justrite™ 12930
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
| ±5% | |
| 2.6 gal. | |
| Polypropylene | |
| White | |
| For storing and dispensing solvents, buffers, reagents and powders. | |
| 10.75 in. | |
| Carboy | |
| 6.4 in. |
| Autoclavable | |
| 10 L | |
| 83 mm | |
| 273 x 163 x 495 mm | |
| 19.5 in. | |
| Polypropylene | |
| Rectangular |