Learn More
Cole-Parmer™ Essentials Polypropylene Cap for E-Vac Aspiration System Vacuum Bottles
Optimize performance of your E-Vac aspiration system
$75.47 - $153.60
Specifications
| Color | Natural |
|---|---|
| Material | Polypropylene |
| Sterility | Non-sterile |
| Thread Size (Metric) | 53B |
| Disposable | Reusable |
Description
- Accessories for E-Vac system designed to collect critical or dangerous liquids
E-Vac aspiration systems available as 04397-10 and 13050-31, sold separately.
M.Vac Jr., polypropylene, Lid, cap
Specifications
| Natural | |
| Non-sterile | |
| Reusable | |
| Not Autoclavable |
| Polypropylene | |
| 53B | |
| E-Vac Aspiration System Vaccum Bottles |