missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate, TCI America™

Catalog No. P19231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19231G Supplier TCI America Supplier No. P19231G

Specifications

CAS 1405-20-5
Color White-Yellow
Content And Storage Refrigerator
Molecular Formula C48H84N16O17S
Format Crystal-Powder at 20°C
MDL Number MFCD00131991
Molecular Weight (g/mol) 1301.571, 1189.36
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O
IUPAC Name 4-amino-N-{1-[(3-amino-1-{[6,9,18-tris(2-aminoethyl)-15-benzyl-3-(2-hydroxyethyl)-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)carbamoyl]-3-hydroxypropyl}-2-formamidobutanamide; sulfuric acid
Product Type Polymyxin B Sulfate
Quantity 1 g
Show More Show Less