missing translation for 'onlineSavingsMsg'
Learn More

Polymedco™ Accessories for SediPlast™ ESR Test: Leveling Plate

Catalog No. 21000122
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21000122 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21000122 Supplier Polymedco™ Supplier No. S1035

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Large Leveling Plate for use with SediPlast ESR Test

Specifications

Sample Type Whole EDTA Blood
Type Large Leveling Plate
For Use With (Application) SediPlast™ ESR test
Quantity Each