missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh

Catalog No. AA43302AB
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43302AB 10 mL
AA43302AD 50 mL
AA43302AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43302AB Supplier Thermo Scientific Chemicals Supplier No. 043302AB

For the separation of nucleotide mixtures | CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays and to pretreat filters to increase the binding of proteins. It can also complex heavy metal ions. An effective permeabilizer of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. It is used as a membrane lubricant, useful in filtering frozen cell membranes and for the separation of nucleotide mixtures. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on PEI cellulose-coated plates. It is also used in separation medium for thin layer chromatography, paper chromatography and Low pressure liquid chromatography. Polyethylenimine on silica gel is used for thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Physical Form Beads
Solubility Information Soluble in water.
Quantity 10 mL
Mesh Size 20 to 40 Mesh
Sensitivity Hygroscopic
Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only