missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Polyethyleneimine hydrochloride, linear, M.W. 40,000
Specifications
Specifications
| CAS | 49553-93-7 |
| Density | 1.05 |
| Molecular Formula | -HN-[CH{2}CH{2}NH-]+n+.HCl |
| MDL Number | MFCD00084427 |
| Quantity | 250 mg |
| Physical Form | Solid |