missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Polyethyleneimine hydrochloride, linear, M.W. 40,000

Catalog No. AA47336MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47336MD 250 mg
AA4733603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47336MD Supplier Thermo Scientific™ Supplier No. 47336MD
Only null left

CAS: 49553-93-7 | -HN-[CH{2}CH{2}NH-]+n+.HCl

Specifications

CAS 49553-93-7
Density 1.05
Molecular Formula -HN-[CH{2}CH{2}NH-]+n+.HCl
MDL Number MFCD00084427
Quantity 250 mg
Physical Form Solid