missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine (ca. 30% in Water), TCI America™

Catalog No. P0381500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0381500G 500 g
P038125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0381500G Supplier TCI America Supplier No. P0381500G
Only null left

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Color Colorless
Physical Form Clear Liquid at 20°C
Quantity 500 g
Chemical Name or Material Polyethyleneimine (ca. 30% in Water)
EINECSNumber (7)-0741
TSCA Yes