missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, branched, M.W. 70,000, 30% w/v aq. soln.

A protein precipitant used to purify proteins | CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

Supplier:  Thermo Scientific Chemicals 04052922

 View more versions of this product

Catalog No. AA4052922


Only null left
Explore available promotions

Description

Description

Acts as a protein precipitant used to purify proteins. It is used as chelating agent and as a scavenger for aldehydes and oxides. Also used in detergents, paper industry, dyes, printing inks and in water treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Colorless to Yellow
M.W. 10,000
9002-98-6
MFCD00084427
Soluble in hot water at a low pH,in methanol,ethanol,or chloroform,but insoluble in cold water,benzene,ethyl ether,and acetone.
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
30% w/v aq. soln.
Polyethyleneimine, branched
Liquid
((C2H5N)zC2H4N)y(C2H5N)x
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
100 g
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
Odorless

Chemical Identifiers

9002-98-6
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TQ7915000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only