Learn More
Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.
CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Supplier: Thermo Scientific Chemicals 04502436
Description
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Colorless to Yellow | |
| Branched, M.W. 50,000 to 100,000 | |
| 9002-98-6 | |
| MFCD00084427 | |
| Soluble in hot water at a low pH,methanol,ethanol,or chloroform. Insoluble in cold water,benzene,ethyl ether and acetone. | |
| NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
| aziridine | |
| 9033 | |
| 30% w/w aq. soln. |
| Polyethyleneimine | |
| Liquid | |
| ((C2H5N)zC2H4N)y(C2H5N)x | |
| ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
| 500 g | |
| *-CCNCCN(-*)CCN-* | |
| 43.07 | |
| CHEBI:30969 | |
| Odorless |
Chemical Identifiers
| 9002-98-6 | |
| 43.07 | |
| NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
| 9033 | |
| aziridine |
| ((C2H5N)zC2H4N)y(C2H5N)x | |
| MFCD00084427 | |
| ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
| CHEBI:30969 | |
| *-CCNCCN(-*)CCN-* |
Safety and Handling
RTECSNumber : TQ7915000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only