Learn More
Kimble™ Polyethylene Snap Cap
Catalog No. S30864E
Change view
Click to view available options
Closure Size:
6
Catalog No. Closure Size
S30864E 6
1 options
Catalog No. S30864E Supplier Kimble™ Supplier No. 28150R6*
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Snap cap tight fit prevents leakage or evaporation.
For Kimax™ Series 28008, 28010, and 28012 volumetric flasks.
Order Info
Sold individually.
Specifications
Closure Color Yellow
Closure Material Polyethylene
For Use With (Equipment) 1000 mL Volumetric Flask
Product Type Snap Cap
Closure Type Snap
Closure Size 6