Learn More
Starplex™ Scientific Polyethylene Plug Caps
Catalog No. 361020454
Change view
Click to view available options
Color:
Green
Lavender
Light Blue
Red
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
361020454 Light Blue
22146428 Green
22146491 White
22146490 Red
22146430 Lavender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361020454 Supplier Starplex™ Scientific Supplier No. V9199
For temporary vial closure
  • Fit 5 and 10mL vials
  • Perform well during specimen processing
  • Nonsterile
  • Not designed for reuse
Specifications
Color Light Blue
Material Polyethylene
Sterility Non-sterile