missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. >5,000,000

Catalog No. AAA1553622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1553622 100 g
AAA1553636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1553622 Supplier Thermo Scientific Chemicals Supplier No. A1553622

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

It is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point ∼175°C
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight >5,000,000
Odor Odorless
Chemical Name or Material Polyethylene oxide
EINECSNumber 500-038-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only