missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 300,000

Catalog No. AA4223736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4223736 500 g
AA4223714 25 g
AA4223722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4223736 Supplier Thermo Scientific Chemicals Supplier No. 04223736

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5 to 7
Physical Form Powder
Melting Point 65°C
Density 1.21 g/mL
Solubility Information Soluble in acetone,alcohol,chloroform,toluene and dichloromethane.
Quantity 500 g
Odor Odorless
Refractive Index 1.4539
Chemical Name or Material Polyethylene oxide, M.W. 300,000
EINECSNumber 500-038-2
RTECSNumber TQ3630000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only