Learn More
Polyethylene oxide, M.W. 100,000
Description
Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| pH | 5 to 7 |
| Physical Form | Powder |
| Melting Point | 65°C |
| Density | 1.4539 g/mL |
| Solubility Information | Soluble in toluene,THF,hot water,DMF,CHCl3Soluble in toluene,terahydrofuran,hot water,dimethylformamide and chloroform. |
| Quantity | 25 g |
| Odor | Odorless |
| Chemical Name or Material | Polyethylene oxide, M.W. 100,000 |
Safety and Handling
| EINECSNumber | 500-038-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only