missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 100,000

Catalog No. AA4223614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4223614 25 g
AA4223622 100 g
AA4223636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4223614 Supplier Thermo Scientific Chemicals Supplier No. 04223614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Physical Form Powder
Melting Point 65°C
Density 1.4539 g/mL
Solubility Information Soluble in toluene,THF,hot water,DMF,CHCl3Soluble in toluene,terahydrofuran,hot water,dimethylformamide and chloroform.
Quantity 25 g
Odor Odorless
Chemical Name or Material Polyethylene oxide, M.W. 100,000
EINECSNumber 500-038-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only