missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol, Spectrum™ Chemical

Catalog No. p-7166147
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612412 500 g Poly Bottle
18612411 2.5 kg Poly Pail
18612410 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612412 Supplier Spectrum Chemical Mfg Cor Supplier No. P3354500GM
Only null left

Chemical Identifiers

CAS 25322-68-3

Specifications

pH 4.5 to 7.5
CAS Max % 1
Quantity 500 g
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.