missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol (PEG), 50% soln.

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Supplier:  Thermo Scientific Chemicals J61495AK

 View more versions of this product

Catalog No. AAJ61495AK


Only null left
Explore available promotions

Description

Description

Polyethylene glycol is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyethylene glycol is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

Solubility
Soluble in water.

Notes
Contains 50% PEG 3350 in 0.1M Lithium Acetate solution, pH 4.9.
Specifications

Specifications

Polyethylene glycol (PEG)
Colorless
MFCD01779601
Soluble in water.
[H]OCCO
62.07
Viscous Liquid
25322-68-3
(C2H4O)n
250 mL
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
50% soln.

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
[H]OCCO
(C2H4O)n
MFCD01779601
ethane-1,2-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only