missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol (PEG), 50% soln.

Catalog No. AAJ61495AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61495AE 100 mL
AAJ61495AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61495AE Supplier Thermo Scientific Chemicals Supplier No. J61495AE

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyethylene glycol is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.

Solubility
Soluble in water.

Notes
Contains 50% PEG 3350 in 0.1M Lithium Acetate solution, pH 4.9.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color Colorless
Quantity 100 mL
Solubility Information Soluble in water.
Concentration or Composition (by Analyte or Components) 50% PEG in 0.1M Lithium Acetate solution
Physical Form Viscous Liquid
Chemical Name or Material Polyethylene glycol (PEG)
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only