missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol, Off-White Powder, MP Biomedicals™
$194.06 - $310.32
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | Carbowax, PEG |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19544501
|
MP Biomedicals Inc
0219544501 |
1 kg |
Each for $194.06
|
|
|||||
|
ICN19544502
|
MP Biomedicals Inc
0219544502 |
2 kg |
Each for $310.32
|
|
|||||
Description
Molecular Biology, ImmunologyChemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| Carbowax, PEG | |
| [H]OCCO |
Specifications
| White | |
| 25322-68-3 | |
| MFCD01779601 | |
| Carbowax, PEG | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 8000 |
| Powder | |
| (C2H4O)n | |
| 1.1-1.2g/mL | |
| 1 kg | |
| [H]OCCO | |
| 62.07 | |
| Polyethylene Glycol |
Safety and Handling
Recommended Storage : Store at room temperature.