missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monooleyl Ether (n=approx. 50), TCI America™

Catalog No. P071625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P071625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P071625G Supplier TCI America Supplier No. P071625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym emulphor, brij 97, oleth-4, emulphor on-870, oleth-20, oleth-23, volpo 20, brij 98, brij 96, emulphor o
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White
Quantity 25 g
Physical Form Flakes
Chemical Name or Material Polyethylene Glycol Monooleyl Ether (n=approx. 50)
EINECSNumber (7)-0097
RTECSNumber RK2975000
TSCA Yes