missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monooleyl Ether (n=approx. 2), TCI America™

Catalog No. P071125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P071125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P071125ML Supplier TCI America Supplier No. P071125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym emulphor, brij 97, oleth-4, emulphor on-870, oleth-20, oleth-23, volpo 20, brij 98, brij 96, emulphor o
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Quantity 25 mL
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol Monooleyl Ether (n=approx. 2)
EINECSNumber (7)-0097
RTECSNumber RK2775000
TSCA Yes