missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethylether, 750

Catalog No. AA4156222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4156222 100 g
AA4156236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4156222 Supplier Thermo Scientific Chemicals Supplier No. 04156222

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol

Polyethylene glycol monomethylether is used as a solvent, excipient, surfactant and dispersing agent. It is also used as a wetting agent and viscosity modifier. It finds application in the cosmetic and pharmaceutical industry due its low toxicity, lubricating property and solubility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
SMILES COCCO

Specifications

Color Colorless to Yellow
Physical Form Low Melting Solid
Melting Point 28°C to 32°C
Density 1.10 g/mL
Solubility Information Slightly miscible with water.
Quantity 100 g
IUPAC Name 2-methoxyethanol
Flash Point >110°C (230°F)
Chemical Name or Material Polyethylene glycol monomethylether, 750
RTECSNumber PE8271000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only