missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethylether, 350

Catalog No. p-7051572
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
AA4156022 100 g
AA4156036 500 g
2 options

Catalog No. AA4156022

Supplier: Thermo Scientific Chemicals 04156022

Only null left

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol

Polyethylene glycol monomethylether is used as enteric release coatings. It is also used for a series of polycarboxylate water reducing agent. It acts as a solvent for brake fluids. Further, it is used in the building materials industry and raw material for cement water reducing agent and strengthening agent. In addition to this, it is used in surfactants, polyester and polyurethane based paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Color Colorless to Yellow
Physical Form Liquid
Density 1.09 g/mL
Solubility Information Partially miscible with water.
Quantity 100 g
Flash Point >110°C (230°F)
Chemical Name or Material Polyethylene glycol monomethylether, 350
RTECSNumber PE8263000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only