missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 18), TCI America™

Catalog No. P070925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P070925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P070925G Supplier TCI America Supplier No. P070925G
Only null left

Chemical Identifiers

CAS 26027-38-3
MDL Number MFCD00148006

Specifications

Color White
Quantity 25 g
Physical Form Wax
Chemical Name or Material Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 18)
EINECSNumber (7)-0172
RTECSNumber RB1366362
TSCA Yes