missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 15), TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Physical Form | Crystals |
| Chemical Name or Material | Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 15) |
Safety and Handling
| EINECSNumber | (7)-0172 |
| RTECSNumber | RB1349544 |
| TSCA | Yes |