missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol methyl ether acrylate, M.W. 5,000

Catalog No. p-7080093
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4638803 1 g
AA4638806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4638803 Supplier Thermo Scientific Chemicals Supplier No. 04638803
Only null left

CAS: 32171-39-4 | (C2H4O)nC4H6O2 | NaN g/mol

Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32171-39-4
Molecular Formula (C2H4O)nC4H6O2
Molecular Weight (g/mol) NaN
MDL Number MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in water
Quantity 1 g
Chemical Name or Material Polyethylene glycol methyl ether acrylate, M.W. 5,000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only