missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Dimethacrylate (n=approx. 4) (stabilized with MEHQ), TCI America™

Catalog No. p-7136729
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T135225G 25 g
T1352500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T135225G Supplier TCI America Supplier No. T135225G
Only null left

Chemical Identifiers

CAS 25852-47-5
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate

Specifications

Color Yellow
Physical Form Clear Liquid at 20°C
Quantity 25 g
Chemical Name or Material Polyethylene Glycol Dimethacrylate (n=approx. 4) (stabilized with MEHQ)
EINECSNumber (7)-1009&(7)-1438
TSCA Yes
Recommended Storage Refrigerator