missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol dimethacrylate, M.W. 8,000

Catalog No. p-7080266
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4698503 1 g
AA4698506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4698503 Supplier Thermo Scientific Chemicals Supplier No. 04698503
Only null left

CAS: 25852-47-5 | CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3

Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25852-47-5
MDL Number MFCD00081877
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate

Specifications

Color White
Physical Form Granular Solid
Molecular Formula CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Solubility Information Soluble in water.
Quantity 1 g
Sensitivity Light and moisture sensitive
Chemical Name or Material Polyethylene glycol dimethacrylate, M.W. 8,000
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only