missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol diacrylate, M.W. 1,000

Catalog No. p-7080116
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46493MD 250 mg
AA4649303 1 g
AA4649306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46493MD Supplier Thermo Scientific Chemicals Supplier No. 046493MD
Only null left

CAS: 26570-48-9 | C5H10O4 | 134.131 g/mol

PEGDA is used in dental compounds, offset inks, varnishes and flexible coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26570-48-9
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00081876
InChI Key FCDZZFLRLVQGEH-UHFFFAOYSA-N
Synonym acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375
PubChem CID 16212859
IUPAC Name ethane-1,2-diol;prop-2-enoic acid
SMILES C=CC(=O)O.C(CO)O

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in THF and chloroform.
Quantity 250 mg
Sensitivity Light sensitive
Chemical Name or Material Polyethylene glycol diacrylate, M.W. 1,000
Hazard Category H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
EINECSNumber 607-960-5
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only