missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 8000, Crystalline Powder, BAKER™, J.T. Baker™

Catalog No. p-5391324
Change view
Click to view available options
Quantity:
1 kg
2 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003647 1 kg Wide Mouth Amber Glass Bottle
02003648 2 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003647 Supplier Avantor Supplier No. U22208
Only null left

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Formula Weight 7000 to 9000 (Average)
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Polyethylene Glycol 8000