missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 6000, Flakes, Ultrapure
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
$180.81 - $180.81
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | PEG 6000 |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ19972A1
|
Thermo Fisher Scientific
J19972A1 |
1 kg | Plastic bottle |
Each for $180.81
|
|
||||
Description
- Differential precipitation of DNA.
- Enhances hybridization rate of nucleic acids.
- Improves efficiency of end-labeling with T4 Polynucleotide Kinase(1,2).
- Used in ligation of blunt ended DNA.
- Precipitates bacteriophage ± particles and plasmid DNA.
- Used in preparing single-stranded M13 DNA.
- Promotes cell fusion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Differential precipitation of DNA
- Enhances hybridization rate of nucleic acids
- Improves efficiency of end-labeling with T4 Polynucleotide Kinase(1,2)
- Used in ligation of blunt ended DNA
- Precipitates bacteriophage ± particles and plasmid DNA
- Used in preparing single-stranded M13 DNA
- Promotes cell fusion
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG 6000 | |
| [H]OCCO |
Specifications
| Flakes | |
| 60°C to 63°C | |
| MFCD01779601 | |
| 1 kg | |
| [H]OCCO | |
| 62.07 | |
| Polyethylene Glycol 6000 |
| 25322-68-3 | |
| (C2H4O)n | |
| PEG 6000 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| Plastic bottle |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
H315-H320
EINECSNumber : 500-038-2
RTECSNumber : TQ3630000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only