missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 6,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Supplier:  Thermo Scientific Chemicals A175410I

 View more versions of this product

Catalog No. AAA175410I


Only null left
Explore available promotions

Description

Description

Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polyethylene glycol 6,000
Powder
60°C to 63°C
MFCD01779601
PEG 6000
5000 g
[H]OCCO
62.07
5 to 7
25322-68-3
(C2H4O)n
14,7568
Soluble in water and aromatic hydrocarbons. Slightly and aliphatic hydrocarbons.
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
Odorless

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
(C2H4O)n
MFCD01779601
PEG 6000
[H]OCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c

H315-H320

EINECSNumber : 203-473-3

RTECSNumber : TQ4100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only