missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 400 Monostearate, Spectrum™ Chemical

Catalog No. 18605330
Change view
Click to view available options
Quantity:
500 g
2.5 kg
18 kg
200 kg
Packaging:
Amber Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605330 18 kg Poly Drum
18605327 500 g Amber Glass Bottle
18605328 2.5 kg Poly-Coated Glass Bottle
18605329 200 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605330 Supplier Spectrum Chemical Mfg Cor Supplier No. P115418KGBL

P1154, 9004-99-3

Spectrum™ Chemical Polyethylene Glycol 400 Monostearate , commonly referred to as PEG 400 MONOSTEARATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9004-99-3
Molecular Formula C20H40O3
Molecular Weight (g/mol) 328.54
InChI Key RFVNOJDQRGSOEL-UHFFFAOYSA-N
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 18 kg
Grade Reagent
Hydroxyl Value 80-100
Moisture 0.03
Packaging Poly Drum