missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 400 Monostearate, Spectrum™ Chemical
Description
Spectrum™ Chemical Polyethylene Glycol 400 Monostearate , commonly referred to as PEG 400 MONOSTEARATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| Acid Value | 2 |
| CAS Max % | 1 |
| Quantity | 200 kg |
| Grade | Reagent |
| Hydroxyl Value | 80-100 |
| Moisture | 0.03 |
| Packaging | Poly Drum |