missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 400 Monostearate, Spectrum™ Chemical
P1154, 9004-99-3
Supplier: Spectrum Chemical Mfg Cor P115425KG
Description
Spectrum™ Chemical Polyethylene Glycol 400 Monostearate , commonly referred to as PEG 400 MONOSTEARATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 9004-99-3 | |
| 1 | |
| 2.5 kg | |
| CCCCCCCCCCCCCCCCCC(=O)OCCO | |
| 328.54 | |
| 80-100 | |
| Poly-Coated Glass Bottle |
| 2 | |
| C20H40O3 | |
| RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
| 2-hydroxyethyl octadecanoate | |
| Reagent | |
| 0.03 |
Chemical Identifiers
| 9004-99-3 | |
| 328.54 | |
| 2-hydroxyethyl octadecanoate |
| C20H40O3 | |
| RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCCO |