missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 400, Liquid, BAKER™, J.T. Baker™

Catalog No. p-5391323
Change view
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003646 500 mL Narrow Mouth Amber Glass Bottle
02003645 4 L Narrow Mouth Amber Glass Bottle
02003644 19 L Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003646 Supplier Avantor Supplier No. U21607
Only null left

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Liquid
Quantity 500 mL
Formula Weight 380 to 420 (Average)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.15kg
Chemical Name or Material Polyethylene Glycol 400