Learn More
Polyethylene glycol 4,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Supplier: Thermo Scientific Chemicals A161510B
Description
Polyethylene glycol is used in gene delivery of protoplast transfection and incubation. It is used in the preparation of extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues. It is useful to mediate DNA transformation into protoplasts derived from rice leaf sheaths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Polyethylene glycol 4,000 | |
| 25322-68-3 | |
| (C2H4O)n | |
| 14,7568 | |
| Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons. | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| Odorless |
| 5 to 7 | |
| 58°C to 62°C | |
| MFCD01779601 | |
| PEG 4000 | |
| 1000 g | |
| [H]OCCO | |
| 62.07 |
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG 4000 | |
| [H]OCCO |
Safety and Handling
RTECSNumber : TQ4050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only