missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 4,000, TCI America™

Catalog No. P0885500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0885500G 500 g
P088525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0885500G Supplier TCI America Supplier No. P0885500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 4,000
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Melting Point 58°C
Quantity 500 g
Chemical Name or Material Polyethylene Glycol 4,000
EINECSNumber (7)-0129
RTECSNumber TQ4050000
TSCA Yes