missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 3350, USP, 97-103%, Spectrum™ Chemical

Catalog No. p-7158017
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605321 12 kg Poly Pail
18605322 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605321 Supplier Spectrum Chemical Mfg Cor Supplier No. PO12512KG
Only null left

PO125, 25322-68-3, H(OCH2CH2)nOH

Spectrum™ Chemical Polyethylene Glycol 3350, USP acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 97 to 103%
Grade USP
Hydroxyl Value 30-38
Ignition Residue 0.001
Packaging Poly Pail
Water 0.01