missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 300, NF, Spectrum™ Chemical
PO108, 25322-68-3, H(OCH2CH2)nOH
$299.07 - $3212.27
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
Description
Spectrum™ Chemical Polyethylene Glycol 300, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| [H]OCCO |
| (C2H4O)n | |
| MFCD01779601 | |
| ethane-1,2-diol |
Specifications
| 4.5 to 7.5 | |
| 1 | |
| MFCD01779601 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| NF | |
| Amber Glass Bottle |
| 25322-68-3 | |
| (C2H4O)n | |
| 500 mL | |
| [H]OCCO | |
| 62.07 | |
| 0.001 | |
| 5.4-6.4 cSt |