missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 12,000

Catalog No. p-7052134
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4263530 250 g
AA42635A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4263530 Supplier Thermo Scientific Chemicals Supplier No. 04263530
Only null left

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Powder
Melting Point 64°C to 65°C
Merck Index 14,7568
Solubility Information Soluble in organic solvents,water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons.
Quantity 250 g
Flash Point >240°C (464°F)
Chemical Name or Material Polyethylene Glycol
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only