missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 1,000

Catalog No. p-3173387
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2213430 250 g
AAB221340B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2213430 Supplier Thermo Scientific Chemicals Supplier No. B2213430
Only null left

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used as an excipent in pharmaceutical products. It is used in the precipitation of proteins as well as in the separation and purification of biomolecules and in the induction of cell hybridization. It acts as a fusing agent to enhance the effect of macrophages on hybridoma; as a vascular agent in preclinical work; as an anti-foaming agent in food and as the gate insulator in an electric double-layer transistor to enhance superconductivity in an insulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Waxy Solid
Melting Point 35°C to 40°C
Density 1.101
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Quantity 250 g
Flash Point 260°C (500°F)
Chemical Name or Material Polyethylene glycol 1000
Hazard Category H227-H303-H315-H320-H333-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
RTECSNumber TQ4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only