Electron Microscopy Sciences POLYETHYLENE GLOVES MEDIUM
Catalog No. 50365595
Low cost ambidextrous powderless/sulfur free gloves with good protection from chemical contamination. Excellent sensitivity. Comes in a dispenser box, 1 3/4 mil thickness
Catalog No.
50365595
Supplier
Electron Microscopy SciencesSupplier No.
71095
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Low cost ambidextrous powderless/sulfur free gloves with good protection from chemical contamination. Excellent sensitivity. Comes in a dispenser box, 1 3/4 mil thickness