missing translation for 'onlineSavingsMsg'
Learn More

Surgical Design™ Polyethylene Dissecting Board

Catalog No. 22079723
Change view
Click to view available options
Size:
13 x 11 in.
23 x 16 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22079723 23 x 16 in.
22079722 13 x 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22079723 Supplier Surgical Design™ Supplier No. 2850216

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Multipurpose polyethylene dissecting board.

  • Will not swell, warp or crack
  • Dimensional scales permit quick sizing and serve as part of permanent record
  • Rubber feet stabilize working surface and help prevent board from slipping
  • White background contrast with specimens for excellent photographic details
  • Feature a spill-containment border

Specifications

Product Type Dissection Board
Autoclavable Autoclavable
Color White
Material Polyethylene
For Use With (Application) Dissection
Quantity 1
Size 23 x 16 in.