missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Sign: WARNING: EAR PROTECTION MUST BE WORN IN THIS AREA. MODERATE HEARING DAMAGE MAY OCCUR.

Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.

$13.62 - $35.04

Specifications

Material Polyester
Shape Rectangular
Displayed Text WARNING: EAR PROTECTION MUST BE WORN IN THIS AREA. MODERATE HEARING DAMAGE MAY OCCUR.
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19111764
View Documents Promotion Details
Brady™
144313
14 x 10 in.
Each for $35.04
Only null left
 
19111763
View Documents Promotion Details
Brady™
144312
10 x 7 in.
Each for $13.62
Only null left
 
Description

Description

  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications

Specifications

Polyester
Rectangular
WARNING: EAR PROTECTION MUST BE WORN IN THIS AREA. MODERATE HEARING DAMAGE MAY OCCUR.
Videos
SDS
Documents

Documents