missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polyester Sign: PINCH POINT. WATCH YOUR HANDS. KEEP HANDS CLEAR. WILL RESULT IN SEVERE INJURY.

Catalog No. 19110806
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19110806 14 x 10 in.
19110805 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19110806 Supplier Brady™ Supplier No. 144035

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Polyester
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text PINCH POINT. WATCH YOUR HANDS. KEEP HANDS CLEAR. WILL RESULT IN SEVERE INJURY.
Color Yellow