missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Sign: DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$18.62 - $18.62
Specifications
| Material | Polyester |
|---|---|
| Shape | Rectangular |
| Displayed Text | DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION. |
| Color | Red on White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19-105-737
|
Brady™
126224 |
7 x 10 in. |
Each for $18.62
|
|
|||||
|
19-105-740
|
Brady™
126227 |
10 x 14 in. |
Each for $18.62
|
|
|||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Polyester | |
| Rectangular | |
| DANGER - FLYING DEBRIS HAZARD. KEEP DOOR CLOSED WHILE IN OPERATION. | |
| Red on White |